accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6064-83-1

IUPAC Name: 2-(phosphonooxy)benzoic acid
Synonyms: fosfosal 磷柳酸
Molecular Weight (g/mol): 218.1
Molecular Formula: C7H7O6P
InChi Key: FFKUDWZICMJVPA-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC=C1OP(O)(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results