accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 607-01-2

IUPAC Name: ethyldiphenylphosphane
Synonyms: phosphine, ethyldiphenyl-
Molecular Weight (g/mol): 214.25
Molecular Formula: C14H15P
InChi Key: WUOIAOOSKMHJOV-UHFFFAOYSA-N
SMILES: CCP(C1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (2)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)