Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyldiphenylphosphine 97.0+%, TCI America™
SDP

Catalog No. E05195G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E05195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E05195G Supplier TCI America Supplier No. E05195G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 607-01-2
Molecular Formula C14H15P
Molecular Weight (g/mol) 214.25
MDL Number MFCD00015170
InChI Key WUOIAOOSKMHJOV-UHFFFAOYSA-N
Synonym ethyldiphenylphosphine, phosphine, ethyldiphenyl, diphenyl ethyl phosphine, diphenylphosphinoethane, diphenylphosphinoethan, ethyl diphenylphosphine, ethyl diphenyl phosphane, acmc-209mlk, ethyl diphenyl phosphine
PubChem CID 69082
IUPAC Name ethyldiphenylphosphane
SMILES CCP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 157°C
Quantity 5 g
Formula Weight 214.25
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyldiphenylphosphine
RTECSNumber SY9242500
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.