CAS RN 607373-46-6
IUPAC Name:
4-[7-(3-aminopyrrolidine-1-carbonyl)-1-ethyl-1H-imidazo[4,5-c]pyridin-2-yl]-1,2,5-oxadiazol-3-amine dihydrochloride
Synonyms:
4-[7-(3-aminopyrrolidine-1-carbonyl)-1-ethylimidazo[4,5-c]pyridin-2-yl]-1,2,5-oxadiazol-3-amine dihydrochloride
Molecular Weight (g/mol):
415.28
Molecular Formula:
C15H20Cl2N8O2
InChi Key:
HSROPTGRSYJPJY-UHFFFAOYNA-N
SMILES:
Cl.Cl.CCN1C(=NC2=CN=CC(C(=O)N3CCC(N)C3)=C12)C1=NON=C1N
1
–
1
of
1
results
1
R&D Systems™ SB 772077B dihydrochloride
CAS: 607373-46-6 Molecular Formula: C15H20Cl2N8O2