accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3CCH3I

CAS RN 60754-76-9

IUPAC Name: benzyltributylazanium iodide
Synonyms: benzyltributylazanium iodide
Molecular Weight (g/mol): 403.39
Molecular Formula: C19H34IN
InChi Key: QVGHRPSUYBFXLH-UHFFFAOYSA-M
SMILES: [I-].CCCC[N+](CCCC)(CCCC)CC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results