Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyltri-n-butylammonium iodide, 97%, Thermo Scientific™

Catalog No. AAB2283714
Change view
Click to view available options
Quantity:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2283714 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2283714 Supplier Thermo Scientific Chemicals Supplier No. B2283714

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 60754-76-9
Molecular Formula C19H34IN
Quantity 25g
MDL Number MFCD00011850
Synonym n-benzyl-n,n-dibutylbutan-1-aminium iodide, benzyltri-n-butylammonium iodide, benzyl tributylammonium iodide, benzyltributylazanium iodide, benzyltributylammonium iodide, tributylbenzyl ammonium iodide, benzyl tributyl ammonium iodide, benzyl tri-n-butylammonium iodide
InChI Key QVGHRPSUYBFXLH-UHFFFAOYSA-M
SMILES CCCC[N+](CCCC)(CCCC)CC1=CC=CC=C1.[I-]
IUPAC Name benzyl(tributyl)azanium;iodide
Molecular Weight (g/mol) 403.392
PubChem CID 16211866
Formula Weight 403.39
Percent Purity 97%
Chemical Name or Material Benzyltri-n-butylammonium iodide
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.