accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 60758-84-1

IUPAC Name: 4-propoxybenzonitrile
Synonyms: 4-propoxybenzonitrile
Molecular Weight (g/mol): 161.2
Molecular Formula: C10H11NO
InChi Key: VYOPNUZYNSNYRA-UHFFFAOYSA-N
SMILES: CCCOC1=CC=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results