accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 60763-78-2

IUPAC Name: 3-(ethylsulfanyl)propane-1,2-diol
Synonyms: 3-(ethylsulfanyl)propane-1,2-diol
Molecular Weight (g/mol): 136.21
Molecular Formula: C5H12O2S
InChi Key: GILAQKILVGHFPX-UHFFFAOYNA-N
SMILES: CCSCC(O)CO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results