accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 60792-79-2

IUPAC Name: dihydrogen 2-(2-aminoethoxy)ethan-1-amine dichloride
Synonyms: dihydrogen 2-(2-aminoethoxy)ethanamine dichloride
Molecular Weight (g/mol): 177.07
Molecular Formula: C4H14Cl2N2O
InChi Key: KTCUXFVANABSPX-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].NCCOCCN
Percent Purity 
  • (1)
  • (1)
  • (2)
Quantity 
  • (3)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)