Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Oxybis(ethylamine) Dihydrochloride 98.0+%, TCI America™
SDP

Catalog No. O02631G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O02631G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O02631G Supplier TCI America Supplier No. O02631G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 60792-79-2
Molecular Formula C4H14Cl2N2O
Molecular Weight (g/mol) 177.07
MDL Number MFCD00012891
InChI Key KTCUXFVANABSPX-UHFFFAOYSA-N
Synonym 2,2'-oxybis ethylamine dihydrochloride, unii-mj9sn78nyt, 2,2'-oxydiethylamine dihydrochloride, mj9sn78nyt, 2,2'-oxybis ethylamine hydrochloride, 2-2-aminoethoxy ethanamine dihydrochloride, 2-aminoethyl ether dihydrochloride, 2,2'-oxydiethanamine dihydrochloride, ethanamine, 2,2'-oxybis-, dihydrochloride
PubChem CID 2829023
IUPAC Name dihydrogen 2-(2-aminoethoxy)ethan-1-amine dichloride
SMILES [H+].[H+].[Cl-].[Cl-].NCCOCCN

Specifications

Melting Point 232°C
Color White
Quantity 1 g
Formula Weight 177.07
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Oxybis(ethylamine) Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.