accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClClClCl

CAS RN 608-93-5

IUPAC Name: 1,2,3,4,5-pentachlorobenzene
Synonyms: pentachlorobenzene QCB pentachloro-benzene C6hcl5 benzene, pentachloro- pentachlorbenzol rcra waste number U183
Molecular Weight (g/mol): 250.32
Molecular Formula: C6HCl5
InChi Key: CEOCDNVZRAIOQZ-UHFFFAOYSA-N
SMILES: ClC1=CC(Cl)=C(Cl)C(Cl)=C1Cl
Molecular Weight (g/mol) 
  • (1)
  • (2)
Grade 
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)