accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 60835-75-8

IUPAC Name: 2,3,5,6,8,9,11,12,14,15-decahydro-1,4,7,10,13,16-benzohexaoxacyclooctadecine-18-carboxylate
Synonyms: 2,3,5,6,8,9,11,12,14,15-decahydro-1,4,7,10,13,16-benzohexaoxacyclooctadecine-18-carboxylate
Molecular Weight (g/mol): 355.36
Molecular Formula: C17H23O8
InChi Key: GFQYJVLOPVAPGJ-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=C2OCCOCCOCCOCCOCCOC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results