Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Carboxybenzo-18-crown 6-Ether 97.0+%, TCI America™
SDP

Catalog No. C17141G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17141G 1 g
C1714100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C17141G Supplier TCI America Supplier No. C17141G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 60835-75-8
Molecular Formula C17H24O8
Molecular Weight (g/mol) 356.371
InChI Key GFQYJVLOPVAPGJ-UHFFFAOYSA-N
Synonym 4'-carboxybenzo-18-crown-6, 4'-carboxybenzo-18-crown 6-ether, 2,3,5,6,8,9,11,12,14,15-decahydro-1,4,7,10,13,16-benzohexaoxacyclooctadecine-18-carboxylic acid, 2,3-4-carboxybenzo-1,4,7,10,13,16-hexaoxacyclooctadec-2-ene, 4-carboxybenzo-18-crown-6, 4/'-carboxybenzo-18-crown 6-ether, benzo-18-crown-6-4'-carboxylic acid, 4 inverted exclamation marka-carboxybenzo-18-crown-6, benzo-18-crown-6-4 inverted exclamation marka-carboxylic acid, 2,5,8,11,14,17-hexaoxabicyclo 16.4.0 docosa-1 18 ,19,21-triene-20-carboxylic acid
PubChem CID 2724805
IUPAC Name 2,5,8,11,14,17-hexaoxabicyclo[16.4.0]docosa-1(18),19,21-triene-20-carboxylic acid
SMILES C1COCCOCCOC2=C(C=CC(=C2)C(=O)O)OCCOCCO1

Specifications

Melting Point 125°C
Quantity 1 g
Formula Weight 356.37
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Carboxybenzo-18-crown 6-Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.