accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 609-14-3

IUPAC Name: ethyl 2-methyl-3-oxobutanoate
Synonyms: ethyl 2-methyl-3-oxobutanoate
Molecular Weight (g/mol): 144.17
Molecular Formula: C7H12O3
InChi Key: FNENWZWNOPCZGK-UHFFFAOYNA-N
SMILES: CCOC(=O)C(C)C(C)=O
Percent Purity 
  • (2)
  • (2)
  • (2)
  • (5)
Quantity 
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (5)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (7)
  • (2)
  • (2)
  • (2)
  • (5)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (5)
  • (2)
  • (2)
  • (6)