Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl 2-methylacetoacetate

Catalog No. E3540025G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E3540025G 25 g
E35400100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E3540025G Supplier Sigma Aldrich Supplier No. E3540025G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Ethyl 2-methylacetoacetate is used as a substrate in the rhenium-catalyzed synthesis of multisubstituted aromatic compounds
  • It can be employed in the synthesis of coumarin derivatives via pechmann condensation
  • It undergoes dehydration to yield conjugated alkynyl and allenyl esters
  • It is also used in the total synthesis of chlorotonil a, yangjinhualine a, (+)- and ()-saudin

Specifications

CAS 609-14-3
Density 1.019 g/mL (at 25°C (literature))
Boiling Point 187°C (lit.)
Molecular Formula C7H12O3
Refractive Index n20/D 1.418 (literature)
Linear Formula CH3COCH(CH3)CO2C2H5
MDL Number MFCD00009164
Quantity 25 g
Synonym Ethyl 2-methyl-3-oxobutanoate
Molecular Weight (g/mol) 144.17
Percent Purity 90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.