accessibility menu, dialog, popup

UserName

Viewing profile as user:
OH(S)(S)OH(R)OH(R)O(S)(R)OH

CAS RN 6090-95-5

IUPAC Name: (1R,2R,3S,4S,5R,6S)-7-oxabicyclo[4.1.0]heptane-2,3,4,5-tetrol
Synonyms: (1R,2R,3S,4S,5R,6S)-7-oxabicyclo[4.1.0]heptane-2,3,4,5-tetrol
Molecular Weight (g/mol): 162.14
Molecular Formula: C6H10O5
InChi Key: ZHMWOVGZCINIHW-FTYOSCRSSA-N
SMILES: O[C@H]1[C@H]2O[C@H]2[C@H](O)[C@@H](O)[C@@H]1O
Quantity 
  • (1)
  • (1)
Form 
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results