Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Conduritol B Epoxide, 98%, MP Biomedicals™

Catalog No. ICN15889325 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15889325 25 mg
ICN15889305 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15889325 Supplier MP Biomedicals, Inc Supplier No. 0215889325
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 6090-95-5 | Formula: C6H10O5 | Weight: 162.141 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6090-95-5
Molecular Formula C6H10O5
Molecular Weight (g/mol) 162.141
InChI Key ZHMWOVGZCINIHW-SCCKRLBHSA-N
Synonym 1r,2r,3s,4r,5r,6s-7-oxabicyclo 4.1.0 heptane-2,3,4,5-tetrol
PubChem CID 25797774
IUPAC Name (1R,2R,3S,4R,5R,6S)-7-oxabicyclo[4.1.0]heptane-2,3,4,5-tetrol
SMILES C1(C(C(C2C(C1O)O2)O)O)O

Specifications

Color White
Density 1.93g/cm3 (L)
Assay Percent Range 0.98
Quantity 25 mg
Formula Weight 162.14
Physical Form Powder
Chemical Name or Material Conduritol B Epoxide
Recommended Storage Store at - 20°C, store desiccated
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.