accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClH3CNCH3NSNCH3CH3

CAS RN 61-73-4

IUPAC Name: 3,7-bis(dimethylamino)-5λ⁴-phenothiazin-5-ylium chloride
Synonyms: methylene blue methylthioninium chloride methylene blue anhydrous methylthioninii chloridum azul de metileno bleu de methylene cloruro de metiltioninio methylenblau chlorure de methylthioninium basic blue 9
Molecular Weight (g/mol): 319.85
Molecular Formula: C16H18ClN3S
InChi Key: CXKWCBBOMKCUKX-UHFFFAOYSA-M
SMILES: [Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (6)
Color 
  • (13)
  • (3)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
  • (11)
  • (1)
  • (3)
Grade 
  • (5)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (5)
  • (5)
  • (2)
  • (1)
  • (2)
  • (4)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (6)
  • (13)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (11)
  • (1)
  • (3)
  • (5)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)