accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(S)NH2OHOCH3

CAS RN 61-90-5

IUPAC Name: (2S)-2-amino-4-methylpentanoic acid
Synonyms: L-leucine
Molecular Weight (g/mol): 131.18
Molecular Formula: C6H13NO2
InChi Key: ROHFNLRQFUQHCH-MLHKIVSYNA-N
SMILES: CC(C)C[C@H](N)C(O)=O
Molecular Weight (g/mol) 
  • (12)
  • (27)
Quantity 
  • (5)
  • (1)
  • (10)
  • (1)
  • (4)
  • (1)
  • (5)
  • (1)
  • (4)
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (5)
  • (4)
  • (11)
  • (5)
  • (3)
Grade 
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)
  • (5)
  • (2)
  • (9)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (19)
  • (17)
  • (1)
  • (4)
  • (1)
  • (1)
  • (6)
  • (13)
  • (3)
  • (8)
  • (4)
  • (3)
  • (11)
  • (12)
  • (27)
  • (5)
  • (1)
  • (10)
  • (1)
  • (4)
  • (1)
  • (5)
  • (1)
  • (4)
  • (1)
  • (3)
  • (1)
  • (5)
  • (4)
  • (11)
  • (5)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)
  • (5)
  • (2)
  • (9)
  • (2)