Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-leucine, 99.42%, MP Biomedicals™

Catalog No. ICN10215805 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10215805 5 kg
ICN10215801 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10215805 Supplier MP Biomedicals, Inc Supplier No. 0210215805
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 61-90-5 | C6H13NO2 | 131.18 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61-90-5
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00002617
InChI Key ROHFNLRQFUQHCH-MLHKIVSYNA-N
Synonym l-leucine, leucine, s-leucine, s-2-amino-4-methylpentanoic acid, h-leu-oh, 2s-2-amino-4-methylpentanoic acid, s-+-leucine, leucin, l-norvaline, 4-methyl, s-2-amino-4-methylvaleric acid
PubChem CID 6106
ChEBI CHEBI:15603
SMILES CC(C)C[C@H](N)C(O)=O

Specifications

Color White
Assay Percent Range 0.9942
Density 1.293g/mL
IUPAC Name 2-amino-4-methylpentanoic acid
Quantity 5 kg
Formula Weight 131.18
Grade Research
Physical Form Small Lustrous Plates or Crystalline Powder
Chemical Name or Material L-Leucine
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.