accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3OHO

CAS RN 610-16-2

IUPAC Name: 2-(dimethylamino)benzoic acid
Synonyms: 2-(dimethylamino)benzoic acid
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: DVVXXHVHGGWWPE-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=CC=C1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results