accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6104-41-2

IUPAC Name: 4-methoxypyrimidine
Synonyms: pyrimidine, 4-methoxy-
Molecular Weight (g/mol): 110.12
Molecular Formula: C5H6N2O
InChi Key: PENVYHXKYYCYML-UHFFFAOYSA-N
SMILES: COC1=CC=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results