Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxypyrimidine 98.0+%, TCI America™
SDP

Catalog No. M23621G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M23621G Supplier TCI America Supplier No. M23621G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6104-41-2
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.116
MDL Number MFCD12405815
InChI Key PENVYHXKYYCYML-UHFFFAOYSA-N
PubChem CID 303830
IUPAC Name 4-methoxypyrimidine
SMILES COC1=NC=NC=C1

Specifications

Color Yellow
Boiling Point 70°C
Quantity 1 g
Formula Weight 110.12
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Methoxypyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.