accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClNHNNNH

CAS RN 6104-71-8

IUPAC Name: 6-chloro-10-(piperazin-1-yl)-2,9-diazatricyclo[9.4.0.0³,⁸]pentadeca-1,3,5,7,10,12,14-heptaene
Synonyms: desmethylclozapine
Molecular Weight (g/mol): 312.8
Molecular Formula: C17H17ClN4
InChi Key: HESZUPIXRNZIOI-UHFFFAOYSA-N
SMILES: ClC1=CC=C2N=C3C=CC=CC3=C(NC2=C1)N1CCNCC1
Color 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)