Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Desmethylclozapine, Thermo Scientific™

Catalog No. AC469191000
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC469191000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC469191000 Supplier Thermo Scientific Chemicals Supplier No. 469191000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 6104-71-8
Molecular Formula C17 H17 Cl N4

Specifications

Quantity 100 mg
Formula Weight 312.8
Physical Form Powder
Color Yellow
Purity ≥97.5%
Chemical Name or Material N-Desmethylclozapine
Hazard Category Exclamation mark
Recommended Storage Normal conditions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.