accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrCH3NO(S)(S)(S)O(R)OO(S)OHH

CAS RN 6106-81-6

IUPAC Name: hydrogen (1S,2R,4S,5S)-7-{[(2S)-3-hydroxy-2-phenylpropanoyl]oxy}-9-methyl-3-oxa-9-azatricyclo[3.3.1.0²,⁴]nonan-9-ium-9-olate bromide
Synonyms: hydrogen (1S,2R,4S,5S)-7-{[(2S)-3-hydroxy-2-phenylpropanoyl]oxy}-9-methyl-3-oxa-9-azatricyclo[3.3.1.0²,⁴]nonan-9-ium-9-olate bromide
Molecular Weight (g/mol): 400.27
Molecular Formula: C17H22BrNO5
InChi Key: MGNNYKWRWHQLCR-XWDDIHSFSA-N
SMILES: [H+].[Br-].C[N+]1([O-])[C@H]2CC(C[C@H]1[C@@H]1O[C@H]21)OC(=O)[C@H](CO)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results