accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONONHOHNO

CAS RN 611-08-5

IUPAC Name: 5-nitro-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: 5-nitrouracil
Molecular Weight (g/mol): 157.09
Molecular Formula: C4H3N3O4
InChi Key: TUARVSWVPPVUGS-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CNC(=O)NC1=O
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)