Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Nitrouracil, 99+%, Thermo Scientific™

Catalog No. AC226580050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC226580050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC226580050 Supplier Thermo Scientific Chemicals Supplier No. 226580050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 611-08-5
Molecular Formula C4H3N3O4
Molecular Weight (g/mol) 157.09
MDL Number MFCD00006021
InChI Key TUARVSWVPPVUGS-UHFFFAOYSA-N
Synonym 5-nitrouracil, 2,4-dihydroxy-5-nitropyrimidine, 5-nitropyrimidine-2,4-diol, uracil, 5-nitro, 5-nitropyrimidine-2,4 1h,3h-dione, 2,4 1h,3h-pyrimidinedione, 5-nitro, 5-nitro-1,2,3,4-tetrahydropyrimidine-2,4-dione, 4-hydroxy-5-nitropyrimidin-2 1h-one, 2,4 1h,3h-pyrimidinedione-5-nitro, 5-nitrouracil 2,4-dihydroxy-5-nitropyrimidine
PubChem CID 69135
ChEBI CHEBI:60763
SMILES [O-][N+](=O)C1=CNC(=O)NC1=O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point >300.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 5 g
Beilstein 24, 320
IUPAC Name 5-nitro-1H-pyrimidine-2,4-dione
Formula Weight 157.08
Percent Purity 99+%
Physical Form Crystalline Powder or Needles
Chemical Name or Material 5-Nitrouracil, p.a.
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-250-4
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.