accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClN

CAS RN 612-57-7

IUPAC Name: 6-chloroquinoline
Synonyms: 6-chloroquinoline
Molecular Weight (g/mol): 163.6
Molecular Formula: C9H6ClN
InChi Key: GKJSZXGYFJBYRQ-UHFFFAOYSA-N
SMILES: ClC1=CC=C2N=CC=CC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results