accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOHOHOOONaNaH2O23

CAS RN 6132-05-4

IUPAC Name: tetrasodium bis(2-(carboxymethyl)-2-hydroxybutanedioate) trihydrate
Synonyms: tetrasodium bis(manganese citrate) trihydrate
Molecular Weight (g/mol): 526.22
Molecular Formula: C12H18Na4O17
InChi Key: HGPVLOQNBSHYEI-UHFFFAOYNA-J
SMILES: O.O.O.[Na+].[Na+].[Na+].[Na+].OC(=O)CC(O)(CC([O-])=O)C([O-])=O.OC(=O)CC(O)(CC([O-])=O)C([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results