accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OClOCH3

CAS RN 6139-83-9

IUPAC Name: 4-chloro-1,1-diethoxybutane
Synonyms: 4-chloro-1,1-diethoxybutane
Molecular Weight (g/mol): 180.67
Molecular Formula: C8H17ClO2
InChi Key: JGGRHRMHOUWCDX-UHFFFAOYSA-N
SMILES: CCOC(CCCCl)OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results