Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorobutyraldehyde Diethyl Acetal 97.0+%, TCI America™
SDP

Catalog No. C27175G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27175G 5 g
C271725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C27175G Supplier TCI America Supplier No. C27175G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6139-83-9
Molecular Formula C8H17ClO2
Molecular Weight (g/mol) 180.672
MDL Number MFCD00671476
InChI Key JGGRHRMHOUWCDX-UHFFFAOYSA-N
Synonym 4-Chloro-1,1-diethoxybutane
PubChem CID 262598
IUPAC Name 4-chloro-1,1-diethoxybutane
SMILES CCOC(CCCCl)OCC

Specifications

Color Yellow
Boiling Point 92°C
Quantity 5 g
UN Number 1993
Formula Weight 180.67
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Chlorobutyraldehyde Diethyl Acetal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.