accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 614-31-3

H2NN

CAS RN 614-31-3

IUPAC Name: 1-benzyl-1-phenylhydrazine
Synonyms: 1-benzyl-1-phenylhydrazine
Molecular Weight (g/mol): 198.27
Molecular Formula: C13H14N2
InChi Key: SQMOOVFBFVTTGF-UHFFFAOYSA-N
SMILES: NN(CC1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results