accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 614-75-5

IUPAC Name: 2-(2-hydroxyphenyl)acetic acid
Synonyms: o-hydroxyphenylacetic acid 2-hpaa o-hydroxy phenylacetic acid
Molecular Weight (g/mol): 152.15
Molecular Formula: C8H8O3
InChi Key: CCVYRRGZDBSHFU-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=CC=C1O
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (4)
  • (4)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (4)