accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClNH2SN

CAS RN 6142-05-8

IUPAC Name: hydrogen 1,3-thiazol-2-amine chloride
Synonyms: hydrogen aminothiazole chloride
Molecular Weight (g/mol): 136.6
Molecular Formula: C3H5ClN2S
InChi Key: WDJXDOCQCALXMV-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC1=NC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results