accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHONFONHO

CAS RN 61422-45-5

IUPAC Name: 5-fluoro-N-hexyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidine-1-carboxamide
Synonyms: carmofur 卡莫氟
Molecular Weight (g/mol): 257.27
Molecular Formula: C11H16FN3O3
InChi Key: AOCCBINRVIKJHY-UHFFFAOYSA-N
SMILES: CCCCCCNC(=O)N1C=C(F)C(=O)NC1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results