Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carmofur 98.0+%, TCI America™
SDP

Catalog No. C266325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C266325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C266325G Supplier TCI America Supplier No. C266325G

Chemical Identifiers

CAS 61422-45-5
Molecular Formula C11H16FN3O3
Molecular Weight (g/mol) 257.27
MDL Number MFCD00866284
InChI Key AOCCBINRVIKJHY-UHFFFAOYSA-N
Synonym 5-Fluoro-1-hexylcarbamoyluracil
PubChem CID 2577
IUPAC Name 5-fluoro-N-hexyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidine-1-carboxamide
SMILES CCCCCCNC(=O)N1C=C(F)C(=O)NC1=O

Specifications

Melting Point 112°C
Color White
Quantity 25 g
UN Number 2811
Formula Weight 257.27
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Carmofur
RTECSNumber UV7714000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.