accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 61437-85-2

IUPAC Name: (2S)-2-(4-amino-2-chloro-5-methylphenyl)-2-(4-chlorophenyl)acetonitrile
Synonyms: (2S)-2-(4-amino-2-chloro-5-methylphenyl)-2-(4-chlorophenyl)acetonitrile
Molecular Weight (g/mol): 291.18
Molecular Formula: C15H12Cl2N2
InChi Key: SCNVPMWFNDBBQS-ZDUSSCGKSA-N
SMILES: CC1=CC([C@@H](C#N)C2=CC=C(Cl)C=C2)=C(Cl)C=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results