accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3COCH3CH3H3C(?)(?)

CAS RN 61440-88-8

IUPAC Name: bis(2,4,6-trimethylphenyl)cycloprop-2-en-1-one
Synonyms: bis(2,4,6-trimethylphenyl)cycloprop-2-en-1-one
Molecular Weight (g/mol): 290.41
Molecular Formula: C21H22O
InChi Key: LXCSCXXFUGZMQN-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(C2=C(C2=O)C2=C(C)C=C(C)C=C2C)C(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results