accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOFFFNH2

CAS RN 61500-87-6

IUPAC Name: methyl 2-amino-4-(trifluoromethyl)benzoate
Synonyms: methyl 2-amino-4-(trifluoromethyl)benzoate
Molecular Weight (g/mol): 219.16
Molecular Formula: C9H8F3NO2
InChi Key: DZICUHOFOOPVFM-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(N)C=C(C=C1)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results