accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 616-14-8

IUPAC Name: 1-iodo-2-methylbutane
Synonyms: 1-iodo-2-methylbutane
Molecular Weight (g/mol): 198.05
Molecular Formula: C5H11I
InChi Key: RHBHXHXNWHTGSO-UHFFFAOYNA-N
SMILES: CCC(C)CI
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results