accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 61607-68-9

IUPAC Name: 1-[2-(dimethylamino)ethyl]-2,5-dihydro-1H-1,2,3,4-tetrazole-5-thione
Synonyms: 1-[2-(dimethylamino)ethyl]-2H-1,2,3,4-tetrazole-5-thione
Molecular Weight (g/mol): 173.24
Molecular Formula: C5H11N5S
InChi Key: ODDAWJGQWOGBCX-UHFFFAOYSA-N
SMILES: CN(C)CCN1NN=NC1=S
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results