accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SNNOHOCl

CAS RN 61727-33-1

IUPAC Name: 5-chloro-2-(methylsulfanyl)pyrimidine-4-carboxylic acid
Synonyms: 5-chloro-2-(methylsulfanyl)pyrimidine-4-carboxylic acid
Molecular Weight (g/mol): 204.63
Molecular Formula: C6H5ClN2O2S
InChi Key: SEPCYCDQJZTPHO-UHFFFAOYSA-N
SMILES: CSC1=NC=C(Cl)C(=N1)C(O)=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)