accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NNH

CAS RN 618-39-3

IUPAC Name: benzenecarboximidamide
Synonyms: benzamidine benzamidine (protonated)
Molecular Weight (g/mol): 120.16
Molecular Formula: C7H8N2
InChi Key: PXXJHWLDUBFPOL-UHFFFAOYSA-N
SMILES: NC(=N)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results