accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 619-33-0

IUPAC Name: 1,1-dichloro-2,2-diethoxyethane
Synonyms: 1,1-dichloro-2,2-diethoxyethane
Molecular Weight (g/mol): 187.06
Molecular Formula: C6H12Cl2O2
InChi Key: CDHLQZJRWKQATP-UHFFFAOYSA-N
SMILES: CCOC(OCC)C(Cl)Cl
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)