Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloroacetal 95.0+%, TCI America™
SDP

Catalog No. D035425ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D035425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D035425ML Supplier TCI America Supplier No. D035425ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 619-33-0
Molecular Formula C6H12Cl2O2
Molecular Weight (g/mol) 187.06
MDL Number MFCD00000849
InChI Key CDHLQZJRWKQATP-UHFFFAOYSA-N
Synonym Dichloroacetaldehyde Diethyl Acetal, 2,2-Dichloro-1,1-diethoxyethane
PubChem CID 12080
IUPAC Name 1,1-dichloro-2,2-diethoxyethane
SMILES CCOC(C(Cl)Cl)OCC

Specifications

Color Yellow
Boiling Point 184°C
Quantity 25 mL
UN Number 1993
Formula Weight 187.06
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Dichloroacetal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.