accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOHO(E)

CAS RN 6192-01-4

IUPAC Name: (2E)-3-ethoxyprop-2-enoic acid
Synonyms: (2E)-3-ethoxyprop-2-enoic acid
Molecular Weight (g/mol): 116.12
Molecular Formula: C5H8O3
InChi Key: SYMAGJYJMLUEQE-ONEGZZNKSA-N
SMILES: CCO\C=C\C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results