accessibility menu, dialog, popup

UserName

Viewing profile as user:
OS

CAS RN 6193-47-1

IUPAC Name: cyclopropyl(thiophen-2-yl)methanone
Synonyms: cyclopropyl(thiophen-2-yl)methanone
Molecular Weight (g/mol): 152.21
Molecular Formula: C8H8OS
InChi Key: PJDFNFSTSCAKPC-UHFFFAOYSA-N
SMILES: O=C(C1CC1)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results