accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 62-38-4

CH3OOHg

CAS RN 62-38-4

IUPAC Name: phenylmercurio acetate
Synonyms: anticon phenylmercuric acetate phenylmercury acetate phenylmercury(II) acetate phenyl mercuric acetate
Molecular Weight (g/mol): 336.74
Molecular Formula: C8H8HgO2
InChi Key: XEBWQGVWTUSTLN-UHFFFAOYSA-M
SMILES: CC(=O)O[Hg]C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)