Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylmercury acetate, 98%

Catalog No. AA3712530
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3712530 250 g
AA3712518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3712530 Supplier Thermo Scientific Chemicals Supplier No. 03712530
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 62-38-4 | C8H8HgO2 | 336.742 g/mol

Phenylmercuric acetate serves as contraceptive gel and foam, injectable solution, skin disinfectant and in cosmetic products such as hair shampoos, mouthwashes and toothpastes. It is likewise used in preservative in canned paint, eye creams and drops. It acts as a catalyst in polyurethane systems. It is also used as a catalyst in the 3M tartan brand polyurethane flexible floors that were common in school gymnasiums. It acts as an antitranspirant, when applied to the plants leaves.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-38-4
Molecular Formula C8H8HgO2
Molecular Weight (g/mol) 336.742
MDL Number MFCD00008691
InChI Key XEBWQGVWTUSTLN-UHFFFAOYSA-M
Synonym phenylmercuric acetate, phenylmercury acetate, acetoxyphenylmercury, anticon, liquiphene, dyanacide, gallotox, hostaquik, kwiksan, meracen
PubChem CID 16682730
ChEBI CHEBI:27684
IUPAC Name acetyloxy(phenyl)mercury
SMILES CC(=O)O[Hg]C1=CC=CC=C1

Specifications

Melting Point 146°C to 153°C
Color White to Yellow
Density 2.4 g/mL
Odor Odorless
Linear Formula C6H5HgOCOCH3
Quantity 250 g
UN Number UN1674
Beilstein 3662930
Merck Index 14,7300
Solubility Information Soluble in alcohol,benzene and glacial acetic acid; Slightly soluble in water.
Formula Weight 336.74
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Phenylmercury Acetate
Show More Show Less
Hazard Category H301-H314-H335-H372-H500
Hazard Statement GHS H Statement
H300-H372-H314-H318
Fatal if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: PHENYLMERCURIC ACETATE
EINECSNumber 200-532-5
RTECSNumber OV6475000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.